Learn More
Medchemexpress LLC Zorevunersen sodium (STK-001 sodium) | 2415330-05-9 | 98.3% | 7197.2 (free acid) | 1 MG
SDP
Catalog No. 5000360397
Encompass_Preferred
Zorevunersen sodium is an antisense oligonucleotide that is intended to increase the level of productive SCN1A mRNA, which in turn increases the expression of the sodium channel Nav1.1 protein. This compound is primarily used for the study of Dravet syndrome.
  • Antisense oligonucleotide for SCN1A mRNA
  • Increases Nav1.1 protein expression
  • High purity
  • Solid, white to off-white appearance
  • Soluble in water at concentrations of 20 mg/mL or greater
  • For research use only
Catalog No. 50-003-60397 Supplier Medchemexpress LLC Supplier No. HY148410A1MG
Zorevunersen sodium is an antisense oligonucleotide that is intended to increase the level of productive SCN1A mRNA, which in turn increases the expression of the sodium channel Nav1.1 protein. This compound is primarily used for the study of Dravet syndrome.
  • Antisense oligonucleotide for SCN1A mRNA
  • Increases Nav1.1 protein expression
  • High purity
  • Solid, white to off-white appearance
  • Soluble in water at concentrations of 20 mg/mL or greater
  • For research use only