Learn More
Medchemexpress LLC HY-18750 10mg Medchemexpress, Zorifertinib CAS:1626387-80-1 Purity:>98%
SDP
Catalog No. 502082083
Encompass_Preferred
Medchemexpress, HY-18750 10mg Zorifertinib CAS:1626387-80-1 Zorifertinib (AZD3759) is a potent, orally active, central nervous system-penetrant, EGFR inhibitor. At Km ATP concentrations, the IC50s are 0.3, 0.2, and 0.2 nM for EGFRwt, EGFRL858R, and EGFRexon 19Del, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2083 Supplier Medchemexpress LLC Supplier No. HY1875010MG
Medchemexpress, HY-18750 10mg Zorifertinib CAS:1626387-80-1 Zorifertinib (AZD3759) is a potent, orally active, central nervous system-penetrant, EGFR inhibitor. At Km ATP concentrations, the IC50s are 0.3, 0.2, and 0.2 nM for EGFRwt, EGFRL858R, and EGFRexon 19Del, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.