Learn More
Medchemexpress LLC Zorifertinib | 1626387-80-1 | 99.4% | 459.90 | 200MG
SDP
Catalog No. 5000386118
Encompass_Preferred
Zorifertinib (AZD3759) is a potent, orally active, and BBB-penetrant EGFR inhibitor. Its IC50s are 0.3, 0.2, and 0.2 nM for EGFR wt, EGFR L858R, and EGFR exon 19Del, respectively, at Km ATP concentrations. This compound induces cancer cell apoptosis and exhibits antitumor activity, making it suitable for research in NSCLC, HCC, and similar conditions.
  • Potent EGFR inhibitor
  • Orally active
  • BBB-penetrant
  • Induces cancer cell apoptosis
  • Exhibits antitumor activity
  • Applicable for NSCLC, HCC, and other research areas
Catalog No. 50-003-86118 Supplier Medchemexpress LLC Supplier No. HY18750200MG
Zorifertinib (AZD3759) is a potent, orally active, and BBB-penetrant EGFR inhibitor. Its IC50s are 0.3, 0.2, and 0.2 nM for EGFR wt, EGFR L858R, and EGFR exon 19Del, respectively, at Km ATP concentrations. This compound induces cancer cell apoptosis and exhibits antitumor activity, making it suitable for research in NSCLC, HCC, and similar conditions.
  • Potent EGFR inhibitor
  • Orally active
  • BBB-penetrant
  • Induces cancer cell apoptosis
  • Exhibits antitumor activity
  • Applicable for NSCLC, HCC, and other research areas