Learn More
Medchemexpress LLC HY-50671 10mg Medchemexpress, Zosuquidar (trihydrochloride) CAS:167465-36-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5067110MG
Encompass_Preferred
Medchemexpress, HY-50671 10mg Zosuquidar (trihydrochloride) CAS:167465-36-3 Zosuquidar trihydrochloride is an inhibitor of P-glycoprotein with a Ki value of 59 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0946