Learn More
TARGETMOL CHEMICALS INC ZOSUQUIDAR TRIHYDROCHLOR 50MG
SDP
Catalog No. 502579652
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zosuquidar trihydrochloride (LY-335979 trihydrochloride) is a potent modulator of P-glycoprotein-mediated multi-drug resistance with Ki of 60 nM. Phase 3. purity: 99%
Catalog No. 50-257-9652 Supplier TARGETMOL CHEMICALS INC Supplier No. T601850MG
Also available in 2 mg 5 mg 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zosuquidar trihydrochloride (LY-335979 trihydrochloride) is a potent modulator of P-glycoprotein-mediated multi-drug resistance with Ki of 60 nM. Phase 3. purity: 99%