Learn More
Cayman Chemical ZosuquIdarhydrochlorIde 10mg
SDP
Catalog No. 502964243
Encompass_Preferred
A modulator of P-gp (Ki = 59 nM); restores the sensitivity of P-gp-expressing cancer cells to vinblastine, doxorubicin, etoposide, and paclitaxel; enhances the distribution of P-gp substrates through the BBB into the brain
Catalog No. 50-296-4243 Supplier Cayman Chemical Supplier No. 2153310 MG
A modulator of P-gp (Ki = 59 nM); restores the sensitivity of P-gp-expressing cancer cells to vinblastine, doxorubicin, etoposide, and paclitaxel; enhances the distribution of P-gp substrates through the BBB into the brain