Learn More
Cayman Chemical Zosuquidar (hydrochloride), Purity: ≥98%, CAS: 167465-36-3, 5 mg
SDP
Catalog No. 502986443
Encompass_Preferred
A modulator of P-gp (Ki = 59 nM); restores the sensitivity of P-gp-expressing cancer cells to vinblastine, doxorubicin, etoposide, and paclitaxel; enhances the distribution of P-gp substrates through the BBB into the brain
Catalog No. 50-298-6443 Supplier Cayman Chemical Supplier No. 215335 MG
A modulator of P-gp (Ki = 59 nM); restores the sensitivity of P-gp-expressing cancer cells to vinblastine, doxorubicin, etoposide, and paclitaxel; enhances the distribution of P-gp substrates through the BBB into the brain