Learn More
Selleck Chemical LLC Zotarolimus
SDP
Catalog No. 5000002180
Encompass_Preferred
Zotarolimus (ABT-578 A 179578) is an analogue of rapamycin and inhibits FKBP-12 binding with IC50 of 2 8 nM
Catalog No. 50-000-02180 Supplier Selleck Chemical LLC Supplier No. S70915MG
Zotarolimus (ABT-578 A 179578) is an analogue of rapamycin and inhibits FKBP-12 binding with IC50 of 2 8 nM