Learn More
Medchemexpress LLC HY-112163 1mg Medchemexpress, Zotatifin CAS:2098191-53-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1121631MG
Encompass_Preferred
Medchemexpress, HY-112163 1mg Zotatifin CAS:2098191-53-6 Zotatifin (eFT226) is a potent, selective, and well-tolerated eIF4A inhibitor. Zotatifin promotes eIF4A binding to specific mRNA sequences with recognition motifs in the 5’-UTRs (IC50=2 nM) and interferes with the assembly of the eIF4F initiation complex. Zotatifin shows robust antiviral effects, it effectively reduces viral infectivity by inhibiting SARS-CoV-2 NP protein biogenesis (IC90=37 nM). Zotatifin induces cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0809