Learn More
Selleck Chemical LLC Zotiraciclib-5mg
SDP
Catalog No. 502389650
Encompass_Preferred
Zotiraciclib (SB1317; TG02) is a novel small molecule potent CDK/JAK2/FLT3 inhibitor, emerged with potent CDK (IC50 against CDKs 1, 2 and 9 = 9, 5 and 3 nM, respectively), FLT3 (IC50 = 19 nM) and JAK2 (IC50 = 19 nM) potency.
Catalog No. 50-238-9650 Supplier Selleck Chemical LLC Supplier No. S70025MG
Zotiraciclib (SB1317; TG02) is a novel small molecule potent CDK/JAK2/FLT3 inhibitor, emerged with potent CDK (IC50 against CDKs 1, 2 and 9 = 9, 5 and 3 nM, respectively), FLT3 (IC50 = 19 nM) and JAK2 (IC50 = 19 nM) potency.