Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zotizalkib | 2648641-36-3 | 98.0% | 447.41 | 50 MG
SDP

Catalog No. 5000348187
Encompass_Preferred

Zotizalkib is a potent, selective, CNS-penetrant, and orally active inhibitor of wild-type ALK and ALK-resistant mutations, such as G1202R and L1196M. It demonstrates strong antitumor activities and is for research use only.

  • Potent, selective, CNS-penetrant, and orally active inhibitor
  • Inhibits wild-type ALK and ALK-resistant mutations
  • Demonstrates strong antitumor activities
  • For research use only
  • Purity: 98.0%
  • Molecular weight: 447.41
  • CAS number: 2648641-36-3

Catalog No. 50-003-48187 Supplier Medchemexpress LLC Supplier No. HY13927950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zotizalkib is a potent, selective, CNS-penetrant, and orally active inhibitor of wild-type ALK and ALK-resistant mutations, such as G1202R and L1196M. It demonstrates strong antitumor activities and is for research use only.

  • Potent, selective, CNS-penetrant, and orally active inhibitor
  • Inhibits wild-type ALK and ALK-resistant mutations
  • Demonstrates strong antitumor activities
  • For research use only
  • Purity: 98.0%
  • Molecular weight: 447.41
  • CAS number: 2648641-36-3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.