Learn More
Medchemexpress LLC Zotizalkib | 2648641-36-3 | 98.0% | C21H20F3N5O3 | 10MG
SDP
Catalog No. 5000260892
Encompass_Preferred
Zotizalkib (TPX-0131) is a potent, CNS-penetrant, orally active inhibitor of anaplastic lymphoma kinase (ALK) designed for research use. It demonstrates low-nanomolar activity against wild-type ALK and a broad spectrum of resistance mutations and has shown antitumor effects in preclinical studies.
  • Potent ALK inhibition (IC50 ≈ 1.4 nM).
  • CNS-penetrant, supporting brain exposure in preclinical models.
  • Active against multiple ALK resistance mutations.
  • High chemical purity (~98.0%).
  • Chemical formula C21H20F3N5O3 and molecular weight 447.41 g/mol.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-002-60892 Supplier Medchemexpress LLC Supplier No. HY13927910MG
Zotizalkib (TPX-0131) is a potent, CNS-penetrant, orally active inhibitor of anaplastic lymphoma kinase (ALK) designed for research use. It demonstrates low-nanomolar activity against wild-type ALK and a broad spectrum of resistance mutations and has shown antitumor effects in preclinical studies.
  • Potent ALK inhibition (IC50 ≈ 1.4 nM).
  • CNS-penetrant, supporting brain exposure in preclinical models.
  • Active against multiple ALK resistance mutations.
  • High chemical purity (~98.0%).
  • Chemical formula C21H20F3N5O3 and molecular weight 447.41 g/mol.
  • Supplied for research use only; not for human or clinical use.