Learn More
Medchemexpress LLC Zovegalisib | 2733573-94-7 | C29H14ClF5N6O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15330610MM/1ML
Encompass_Preferred
Zovegalisib (RLY-2608) is an orally active, first-in-class, allosteric, mutant-selective inhibitor of PI3Kα with anti-tumor activity. It inhibits tumor growth in PIK3CA-mutant xenograft mouse models with minimal impact on insulin levels. This compound is for research use only and not sold to patients.
  • First-in-class, allosteric, mutant-selective inhibitor of PI3Kα
  • Exhibits anti-tumor activity
  • Inhibits tumor growth in PIK3CA-mutant xenograft mice models
  • Minimal impact on insulin levels
  • High purity (98.80%)
Catalog No. 50-003-73481