Learn More
AdipoGen Zoxamide
SDP
Catalog No. 502559867
Encompass_Preferred
Chemical. CAS 156052-68-5. Formula C14H16Cl3NO2. MW 336.6. Synthetic. Fungicide. Primarily an oomycete fungicide although it has some activity against ascomycetes and basidiomycetes. The product acts by disruption of the microtubule structure and inhibition of cell division. The mechanism for this activity is through the covalent binding of the active ingredient to beta-tubulin in the cytoskeleton. Scanning microscopy studies showed zoxamide action against blight on potato crops begins after germination, achieving inhibition of hyphal development and sporangia production. This novel mode of action makes zoxamide an important new tool in antiresistance strategies. Can be used as a reference compound.
Catalog No. 50-255-9867 Supplier AdipoGen Supplier No. CDXZ0015M100
Chemical. CAS 156052-68-5. Formula C14H16Cl3NO2. MW 336.6. Synthetic. Fungicide. Primarily an oomycete fungicide although it has some activity against ascomycetes and basidiomycetes. The product acts by disruption of the microtubule structure and inhibition of cell division. The mechanism for this activity is through the covalent binding of the active ingredient to beta-tubulin in the cytoskeleton. Scanning microscopy studies showed zoxamide action against blight on potato crops begins after germination, achieving inhibition of hyphal development and sporangia production. This novel mode of action makes zoxamide an important new tool in antiresistance strategies. Can be used as a reference compound.