Learn More
Aviva Systems Biology Corp ZP1 Antibody - middle region (ARP55991_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55991_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZP1. Alternate names: MGC87693ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61687