Learn More
Aviva Systems Biology Corp ZP2 Antibody - C-terminal region (ARP45470_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45470_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZP2. Alternate names: ZPA, Zp-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-53448