Learn More
Aviva Systems Biology Corp ZP4 Antibody - N-terminal region (ARP49639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZP4. Alternate names: ZBP, ZP1, ZPB, Zp-4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60133