Learn More
Aviva Systems Biology Corp ZPBP2 Antibody - N-terminal region (ARP53476_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZPBP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZPBP2. Alternate names: MGC41930, ZPBPLApplicationWB. Species reactivity: Human
Catalog No. 50-285-841