Learn More
Aviva Systems Biology Corp ZPBP2 Antibody - middle region (ARP53477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZPBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZPBP2. Alternate names: MGC41930, ZPBPLApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63035