Learn More
Cayman Chemical ZPheTyr tBudIazomethylket 10mg
SDP
Catalog No. 502937427
Encompass_Preferred
An inhibitor of cathepsin L (Kinact = 200,000 M-1s-1); selective for cathepsin L over cathepsin S and cathepsin B (Kinacts = 30 and 10.3 M-1s-1, respectively); active against bloodstream forms of T. brucei brucei clone 427-221a in vitro (EC50 = 126 μM)
Catalog No. 50-293-7427 Supplier Cayman Chemical Supplier No. 2787710 MG
An inhibitor of cathepsin L (Kinact = 200,000 M-1s-1); selective for cathepsin L over cathepsin S and cathepsin B (Kinacts = 30 and 10.3 M-1s-1, respectively); active against bloodstream forms of T. brucei brucei clone 427-221a in vitro (EC50 = 126 μM)