Learn More
Trajan Scientific ZPure H2O filter, 1/4" brass fittings, with Mol Sieve 3A
SDP
Catalog No. 502724275
Encompass_Preferred
ZPure H2O filter, 1/4" brass fittings, with Mol Sieve 3A
Catalog No. 50-272-4275 Supplier Trajan Scientific Supplier No. 203011B
ZPure H2O filter, 1/4" brass fittings, with Mol Sieve 3A