Learn More
Trajan Scientific ZPure O2 / H2O filter, 1/4" stainless steel compression fittings
SDP
Catalog No. 502724486
Encompass_Preferred
ZPure O2 / H2O filter, 1/4" stainless steel compression fittings
Catalog No. 50-272-4486 Supplier Trajan Scientific Supplier No. 202296SS
ZPure O2 / H2O filter, 1/4" stainless steel compression fittings