Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, ZQ, EMD1000, Quattro Micro MS, Annual, No Chemical Kit
Catalog No. 50791591 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This performance maintenance kit is designed for ZQ, EMD1000, and Quattro Micro Mass Spectrometers. It contains all the necessary quality parts to ensure the instrument operates at peak performance when replaced annually. The kit includes various components such as an ESI probe kit, sample cone, and ultragrade oil. Note that a chemical kit, essential for performance testing, is not included.
  • Designed for ZQ, EMD1000, and Quattro Micro Mass Spectrometers.
  • Ensures instrument operates at peak performance.
  • Recommended for annual replacement.
  • Includes an ESI probe kit, sample cone, lapping film, inner API source kit, outer API source kit, oil mist filter element, powder-free gloves, source heater assembly, fused silica tube, 2 L of ultragrade 19 oil, and an odor element.
  • Chemical kit not included, which is required for performance testing.
Catalog No. 50-791-591 Supplier Waters Corp Supplier No. 201000213
Add to Cart
Add to Cart
Request a Quote
This performance maintenance kit is designed for ZQ, EMD1000, and Quattro Micro Mass Spectrometers. It contains all the necessary quality parts to ensure the instrument operates at peak performance when replaced annually. The kit includes various components such as an ESI probe kit, sample cone, and ultragrade oil. Note that a chemical kit, essential for performance testing, is not included.
  • Designed for ZQ, EMD1000, and Quattro Micro Mass Spectrometers.
  • Ensures instrument operates at peak performance.
  • Recommended for annual replacement.
  • Includes an ESI probe kit, sample cone, lapping film, inner API source kit, outer API source kit, oil mist filter element, powder-free gloves, source heater assembly, fused silica tube, 2 L of ultragrade 19 oil, and an odor element.
  • Chemical kit not included, which is required for performance testing.