Learn More
Zymo Research Corporation ZR 1 kb DNA Marker™ (Ready-to-load) (50 μg/ 600 μl)
SDP
Catalog No. 501251649
Encompass_Preferred
The 1 kb DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 10 defined fragments that range in size from 0.5 - 10 kb. The intensity of the 3 kb band is increased as an identifier. All fragments have 4-base, 5 overhangs.
Catalog No. 50-125-1649 Supplier Zymo Research Corporation Supplier No. M500650
The 1 kb DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 10 defined fragments that range in size from 0.5 - 10 kb. The intensity of the 3 kb band is increased as an identifier. All fragments have 4-base, 5 overhangs.