Learn More
Zymo Research Corporation ZR 100 bp DNA Marker™ (200 μg / 400 μl)
SDP
Catalog No. 502037209
Encompass_Preferred
The 100 bp DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 12 defined fragments that range in size from 100 - 1500 bp.The intensity of the 500 and 1000 bp band is increased as an identifier. All fragments have 4-base, 5 overhangs.
Catalog No. 50-203-7209 Supplier Zymo Research Corporation Supplier No. M5002200
The 100 bp DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 12 defined fragments that range in size from 100 - 1500 bp.The intensity of the 500 and 1000 bp band is increased as an identifier. All fragments have 4-base, 5 overhangs.