Learn More
Zymo Research Corporation ZR 50 bp DNA Marker™ (Ready-to-load) (50 μg/ 600 μl)
SDP
Catalog No. 501251647
Encompass_Preferred
The 50 bp DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 12 defined fragments that range in size from 50 - 200 bp. The intensity of the 300 bp band is increased as an identifier. All fragments have 4-base, 5 overhangs.
Catalog No. 50-125-1647 Supplier Zymo Research Corporation Supplier No. M500450
The 50 bp DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 12 defined fragments that range in size from 50 - 200 bp. The intensity of the 300 bp band is increased as an identifier. All fragments have 4-base, 5 overhangs.