Learn More
Zymo Research Corporation ZR-96 BashingBead™ Lysis Rack (2 mm) (2 racks)
SDP
Catalog No. 501251734
Encompass_Preferred
Tubes are in a 96-well rack with caps and a cover for high throughput processing. The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tough-to-lyse biological samples. These beads are ideal for microbes and fungi in soil, feces, sludge, etc.
Catalog No. 50-125-1734 Supplier Zymo Research Corporation Supplier No. S6002962
Tubes are in a 96-well rack with caps and a cover for high throughput processing. The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tough-to-lyse biological samples. These beads are ideal for microbes and fungi in soil, feces, sludge, etc.