Learn More
Zymo Research Corporation ZR-96 ChIP DNA Clean & Concentrator™ (4x96 preps)
SDP
Catalog No. 502037139
Encompass_Preferred
The ZR-96 Chromatin Immunoprecipitation (ChIP) DNA Clean & Concentrator provides a hassle-free method for the rapid, high-throughput purification, and concentration of high-quality DNA from any step in a standard ChIP protocol. This product is well suited for samples that have undergone reverse cross-linking and Proteinase K or RNase A digestion as well as reverse cross-linked samples eluted from chromatin-antibody-bead complexes. Additionally, this product may also be used to purify DNA from PCR and other enzymatic reactions. DNA purified using the ZR-96 ChIP DNA Clean & Concentrator™ is suitable for PCR, Next-Gen sequencing (ChIP-Seq), arrays, as well as other molecular applications.
Catalog No. 50-203-7139 Supplier Zymo Research Corporation Supplier No. D5207
The ZR-96 Chromatin Immunoprecipitation (ChIP) DNA Clean & Concentrator provides a hassle-free method for the rapid, high-throughput purification, and concentration of high-quality DNA from any step in a standard ChIP protocol. This product is well suited for samples that have undergone reverse cross-linking and Proteinase K or RNase A digestion as well as reverse cross-linked samples eluted from chromatin-antibody-bead complexes. Additionally, this product may also be used to purify DNA from PCR and other enzymatic reactions. DNA purified using the ZR-96 ChIP DNA Clean & Concentrator™ is suitable for PCR, Next-Gen sequencing (ChIP-Seq), arrays, as well as other molecular applications.