Learn More
Research Products International Corp ZR-96 DNA Clean & Concentrator-5 Kit, 2 x 96 Preps
SDP
Catalog No. 501977400
Encompass
The ZR-96 DNA Clean & Concentrator-5 products provide purification of up to 5 µg DNA per well from PCR, endonuclease digestions, DNA modification reactions, isotope/fluorescence labeling reactions, etc. The product facilitates the removal of DNA polymerases, modifying enzymes, RNA polymerases, ligases, kinases, nucleases, phosphatases, and restriction endonucleases, as well as free dNTPs and their analogs including radiolabeled and fluorescent derivatives. Eluted DNA is suitable for PCR, arrays, ligation, sequencing, etc.
Clean and concentrate up to 5 μg DNA (per well) with ≥ 10 μl elution volume in as little as 15 minutes with 0 μl wash residue carryover.
Deep-well filtration plate designs allow DNA to be eluted at high concentrations into minimal volumes of water or TE buffer.
Catalog No. 50-197-7400 Supplier Research Products International Corp Supplier No. ZD4023
The ZR-96 DNA Clean & Concentrator-5 products provide purification of up to 5 µg DNA per well from PCR, endonuclease digestions, DNA modification reactions, isotope/fluorescence labeling reactions, etc. The product facilitates the removal of DNA polymerases, modifying enzymes, RNA polymerases, ligases, kinases, nucleases, phosphatases, and restriction endonucleases, as well as free dNTPs and their analogs including radiolabeled and fluorescent derivatives. Eluted DNA is suitable for PCR, arrays, ligation, sequencing, etc.
Clean and concentrate up to 5 μg DNA (per well) with ≥ 10 μl elution volume in as little as 15 minutes with 0 μl wash residue carryover.
Deep-well filtration plate designs allow DNA to be eluted at high concentrations into minimal volumes of water or TE buffer.