Learn More
Zymo Research Corporation ZR-96 DNA Clean-up Kit™ Kit (4 x 96 Preps)
SDP
Catalog No. NC9074001
Encompass_Preferred
Simply add the specially formulated DNA Binding Buffer to your samples and transfer to the wells of the supplied SiliconA™ Plate. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield highquality, purified DNA in just minutes. DNA purified using the ZR-96 DNA Clean-up Kit™ is suitable for nucleotide sequencing, microarray analysis, PCR, nucleotide blotting, and restriction endonuclease digestion procedures. The ZR-96 DNA Clean-up Kit™ employs a single-buffer system that allows for efficient DNA adsorption of up to 96 samples onto the matrix of the supplied Silicon-A™ Plate. The DNA is washed twice then eluted with a small volume of water into a 96-well Elution Plate. The entire DNA purification/concentration procedure typically takes about 20 minutes.
Catalog No. NC9074001 Supplier Zymo Research Corporation Supplier No. D4018
Simply add the specially formulated DNA Binding Buffer to your samples and transfer to the wells of the supplied SiliconA™ Plate. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield highquality, purified DNA in just minutes. DNA purified using the ZR-96 DNA Clean-up Kit™ is suitable for nucleotide sequencing, microarray analysis, PCR, nucleotide blotting, and restriction endonuclease digestion procedures. The ZR-96 DNA Clean-up Kit™ employs a single-buffer system that allows for efficient DNA adsorption of up to 96 samples onto the matrix of the supplied Silicon-A™ Plate. The DNA is washed twice then eluted with a small volume of water into a 96-well Elution Plate. The entire DNA purification/concentration procedure typically takes about 20 minutes.