Learn More
Zymo Research Corporation ZR-96 DNA Clean & Concentrator™-5 Kit (4 x 96 Preps)
SDP
Catalog No. NC9940044
Encompass_Preferred
The ZR-96 DNA Clean & Concentrator-5 (ZR-96 DCC-5) can be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer to the wells of the supplied Zymo-Spin™ I-96 Plate. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin technology to yield DNA that is free of salts and contaminants in just minutes. DNA purified using the ZR-96 DCC-5 is suitable for nucleotide sequencing, microarray analysis, PCR, and restriction endonuclease digestion procedures.
Catalog No. NC9940044 Supplier Zymo Research Corporation Supplier No. D4024
The ZR-96 DNA Clean & Concentrator-5 (ZR-96 DCC-5) can be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer to the wells of the supplied Zymo-Spin™ I-96 Plate. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin technology to yield DNA that is free of salts and contaminants in just minutes. DNA purified using the ZR-96 DCC-5 is suitable for nucleotide sequencing, microarray analysis, PCR, and restriction endonuclease digestion procedures.