Learn More
Zymo Research Corporation Quick-DNA™ 96 Kit™ (2 x 96 Preps)
SDP
Supplier:  Zymo Research Corporation D3010
Encompass_Preferred
This product has been optimized for maximal recovery of ultra-pure DNA without RNA contamination and is compatible with whole blood (fresh or stored), serum, plasma, buffy coat, buccal cells, cells from culture, and many biological liquid samples.For processing, simply add the specially formulated Genomic Lysis Buffer to the samples, vortex, and transfer the mixtures to the wells of the supplied SiliconA™ Plate. There is no need for organic denaturants or Proteinase K digestion because of the unique chemistries featured in the kit. Instead, the product yields high-quality, purified DNA in just minutes (see below). PCR inhibitors are effectively removed during the purification process. DNA purified using the Quick-DNA™ 96 Kit is suitable for PCR, nucleotide blotting, DNA sequencing, restriction endonuclease digestion, bisulfite conversion/methylation analysis, and other downstream applications.
Catalog No. 50-444-137