Learn More
Research Products International Corp ZR-96 Quick-RNA Kit, 4 x 96 Preps
SDP
Catalog No. 501976952
Encompass
The Quick-RNA 96 Kit is one of the most innovative RNA isolation kits available, designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell (up to 106) and tissue samples (up to 5 mg). The procedure combines a unique buffer system with Zymo-Spin plate technology to yield high quality total RNA (including small RNAs ~17-200 nt) in about 30 minutes. The procedure is simple: Add the provided RNA Lysis Buffer to a sample, then purify the RNA using the provided Silicon-A Plate. The result is highly-concentrated, DNA-free RNA that is suitable for subsequent RNA-based methods including RT-PCR, hybridization, sequencing etc.
Broad Range: Extract total RNA (including small/micro RNAs) from any sample.
DNA-Free: Genomic DNA removal column and DNase I included.
NGS-Ready: RNA is ready for all downstream applications including Next-Gen Sequencing, RT-qPCR, hybridization, etc.
Catalog No. 50-197-6952 Supplier Research Products International Corp Supplier No. ZR1053
The Quick-RNA 96 Kit is one of the most innovative RNA isolation kits available, designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell (up to 106) and tissue samples (up to 5 mg). The procedure combines a unique buffer system with Zymo-Spin plate technology to yield high quality total RNA (including small RNAs ~17-200 nt) in about 30 minutes. The procedure is simple: Add the provided RNA Lysis Buffer to a sample, then purify the RNA using the provided Silicon-A Plate. The result is highly-concentrated, DNA-free RNA that is suitable for subsequent RNA-based methods including RT-PCR, hybridization, sequencing etc.
Broad Range: Extract total RNA (including small/micro RNAs) from any sample.
DNA-Free: Genomic DNA removal column and DNase I included.
NGS-Ready: RNA is ready for all downstream applications including Next-Gen Sequencing, RT-qPCR, hybridization, etc.