Learn More
Zymo Research Corporation ZR BAC DNA Miniprep Kit, 100 preps
SDP
Catalog No. 50444258
Encompass_Preferred
The ZR BAC DNA Miniprep Kit efficiently isolates BAC plasmid DNA or other large plasmids (e.g., PAC) from E. Coli using a procedure that is simple, rapid, user-friendly, and reliable. It features a modified alkaline lysis protocol with color-coded reagents that allow easy visualization of complete bacterial cell lysis and neutralization. The innovative Zymo-Spin™ IC-XL columns are optimized for high yield, endotoxin-free plasmid DNA recovery. BAC DNA purified using the ZR BAC DNA Miniprep Kit is ideal for sequencing, PCR, endonuclease digestion, etc.
Catalog No. 50-444-258 Supplier Zymo Research Corporation Supplier No. D4049
The ZR BAC DNA Miniprep Kit efficiently isolates BAC plasmid DNA or other large plasmids (e.g., PAC) from E. Coli using a procedure that is simple, rapid, user-friendly, and reliable. It features a modified alkaline lysis protocol with color-coded reagents that allow easy visualization of complete bacterial cell lysis and neutralization. The innovative Zymo-Spin™ IC-XL columns are optimized for high yield, endotoxin-free plasmid DNA recovery. BAC DNA purified using the ZR BAC DNA Miniprep Kit is ideal for sequencing, PCR, endonuclease digestion, etc.