Learn More
Zymo Research Corporation ZR BashingBead Lysis Tubes (0.1 & 0.5 mm), 50 tubes
SDP
Catalog No. nc1099697
Encompass_Preferred
The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tough-to-lyse biological samples. These beads are ideal for tough-to-lyse bacteria, yeast, fungi and algae.
Catalog No. NC1099697 Supplier Zymo Research Corporation Supplier No. S601250
The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tough-to-lyse biological samples. These beads are ideal for tough-to-lyse bacteria, yeast, fungi and algae.