Learn More
Zymo Research Corporation ZR BashingBead™ Lysis Tubes (2 mm) (50 tubes)
SDP
Catalog No. 50444640
Encompass_Preferred
Each impact resistant 2 ml tube contains 0.7 ml dry volume 2.0 mm ZR BashingBead™ lysis matrix. The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tough-to-lyse biological samples. These beads are ideal for tissues, insects, plant material, etc.
Catalog No. 50-444-640 Supplier Zymo Research Corporation Supplier No. S600350
Each impact resistant 2 ml tube contains 0.7 ml dry volume 2.0 mm ZR BashingBead™ lysis matrix. The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tough-to-lyse biological samples. These beads are ideal for tissues, insects, plant material, etc.