Learn More
Zymo Research Corporation ZR BashingBead™ Lysis Tubes (0.1 mm & 2.0 mm); 50 pack
SDP
Catalog No. 502037282
Encompass_Preferred
The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting microbial containing vectors such as ticks, fleas, mosquitoes, and tissues. Used in the Quick-DNA/RNA Pathogen Miniprep kit (Cat. #R1042).
Catalog No. 50-203-7282 Supplier Zymo Research Corporation Supplier No. S601450
The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting microbial containing vectors such as ticks, fleas, mosquitoes, and tissues. Used in the Quick-DNA/RNA Pathogen Miniprep kit (Cat. #R1042).