Learn More
Zymo Research Corporation ZR DNA Sequencing Clean-up Kit™ (200 Preps) w/ Zymo Spin IB
SDP
Catalog No. 50444266
Encompass_Preferred
The ZR DNA Sequencing Clean-Up Kits provide a simple and rapid (2 & 10 minute) method for removal of post-cycle sequencing reaction contaminants (i.e., unincorporated fluorescent dyes, residual salts, dNTPs, primers, and enzymes) from DNA extension products. These contaminants can often interfere with the quality and signal strength of sequencing data, including dye peaks or "dye blobs" which may obscure portions of the sequencing chromatogram and interfere with base-calling accuracy of sequencing analysis software. DNA is eluted with a small volume of water or loading dye containing formamide.
Catalog No. 50-444-266 Supplier Zymo Research Corporation Supplier No. D4051
The ZR DNA Sequencing Clean-Up Kits provide a simple and rapid (2 & 10 minute) method for removal of post-cycle sequencing reaction contaminants (i.e., unincorporated fluorescent dyes, residual salts, dNTPs, primers, and enzymes) from DNA extension products. These contaminants can often interfere with the quality and signal strength of sequencing data, including dye peaks or "dye blobs" which may obscure portions of the sequencing chromatogram and interfere with base-calling accuracy of sequencing analysis software. DNA is eluted with a small volume of water or loading dye containing formamide.