Learn More
Zymo Research Corporation ZR DNA Sequencing Clean-up Kit™ (50 Preps) w/ Zymo Spin IB
SDP
Catalog No. 50444259
Encompass_Preferred
The ZR DNA Sequencing Clean-Up Kits provide a simple and rapid (2 & 10 minute) method for removal of post-cycle sequencing reaction contaminants (i.e., unincorporated fluorescent dyes, residual salts, dNTPs, primers, and enzymes) from DNA extension products. These contaminants can often interfere with the quality and signal strength of sequencing data, including dye peaks or "dye blobs" which may obscure portions of the sequencing chromatogram and interfere with base-calling accuracy of sequencing analysis software. DNA is eluted with a small volume of water or loading dye containing formamide.
Catalog No. 50-444-259 Supplier Zymo Research Corporation Supplier No. D4050
The ZR DNA Sequencing Clean-Up Kits provide a simple and rapid (2 & 10 minute) method for removal of post-cycle sequencing reaction contaminants (i.e., unincorporated fluorescent dyes, residual salts, dNTPs, primers, and enzymes) from DNA extension products. These contaminants can often interfere with the quality and signal strength of sequencing data, including dye peaks or "dye blobs" which may obscure portions of the sequencing chromatogram and interfere with base-calling accuracy of sequencing analysis software. DNA is eluted with a small volume of water or loading dye containing formamide.