Learn More
Zymo Research Corporation Quick-DNA Fecal/Soil Microbe MicroPrep™ Kit (50 Preps)
SDP
Catalog No. 50444459
Encompass_Preferred
The kit can be used to successfully isolate DNA from tough-to-lyse Gram positive and negative bacteria, fungi, algae, protozoa, etc. that inhabit fecal and soil samples. The procedure is easy and can be completed in as little as 15 minutes: fecal samples (50 mg each) or soil samples (250 mg each) are added directly to a ZR BashingBead™ Lysis Tube (0.1 & 0.5 mm) and rapidly and efficiently lysed by bead beating without the use of organic denaturants or proteinases. The DNA is then isolated using our Zymo-Spin™ Technology, which is subsequently filtered to remove humic acids/polyphenols that inhibit PCR. The DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, etc.
Catalog No. 50-444-459 Supplier Zymo Research Corporation Supplier No. D6012
The kit can be used to successfully isolate DNA from tough-to-lyse Gram positive and negative bacteria, fungi, algae, protozoa, etc. that inhabit fecal and soil samples. The procedure is easy and can be completed in as little as 15 minutes: fecal samples (50 mg each) or soil samples (250 mg each) are added directly to a ZR BashingBead™ Lysis Tube (0.1 & 0.5 mm) and rapidly and efficiently lysed by bead beating without the use of organic denaturants or proteinases. The DNA is then isolated using our Zymo-Spin™ Technology, which is subsequently filtered to remove humic acids/polyphenols that inhibit PCR. The DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, etc.