Learn More
Zymo Research Corporation Quick-DNA Fungal/Bacterial Midiprep Kit (25 Preps)
SDP
Catalog No. 50444474
Encompass_Preferred
The procedure is easy and can be completed in as little as 20 minutes: fungal and/or bacterial samples are added directly to a ZR BashingBead™ Lysis/Filtration Tube and rapidly and efficiently lysed by bead beating without using organic denaturants or proteinases. The DNA is isolated and purified using our FastSpin column technology and is ideal for downstream molecular-based applications including PCR, array, etc.
Catalog No. 50-444-474 Supplier Zymo Research Corporation Supplier No. D6105
The procedure is easy and can be completed in as little as 20 minutes: fungal and/or bacterial samples are added directly to a ZR BashingBead™ Lysis/Filtration Tube and rapidly and efficiently lysed by bead beating without using organic denaturants or proteinases. The DNA is isolated and purified using our FastSpin column technology and is ideal for downstream molecular-based applications including PCR, array, etc.