Learn More
Zymo Research Corporation Quick-RNA Fungal/Bacterial MicroPrep™ Kit (50 Preps)
SDP
Catalog No. 50444616
Encompass_Preferred
Ultra-high density ZR BashingBeads™ are included for sample homogenization coupled with a robust buffer system, delivering total RNA (including small/microRNAs). The Zymo-Spin™ IIICG Column allows for high-capacity DNA elimination and the subsequent Zymo-Spin™ IC Column efficiently binds total RNA.RNA is eluted in as little as 6 l and is suitable for subsequent procedures including RT-qPCR.
Catalog No. 50-444-616 Supplier Zymo Research Corporation Supplier No. R2010
Ultra-high density ZR BashingBeads™ are included for sample homogenization coupled with a robust buffer system, delivering total RNA (including small/microRNAs). The Zymo-Spin™ IIICG Column allows for high-capacity DNA elimination and the subsequent Zymo-Spin™ IC Column efficiently binds total RNA.RNA is eluted in as little as 6 l and is suitable for subsequent procedures including RT-qPCR.