Learn More
Zymo Research Corporation Quick-RNA Fungal/Bacterial MiniPrep™ Kit (50 Preps)
SDP
Catalog No. 50444617
Encompass_Preferred
Ultra-high density ZR BashingBeads™ are included for sample homogenization coupled with a robust buffer system, delivering total RNA (including small/microRNAs).The Zymo-Spin™ IIICG Column allows for high-capacity DNA elimination and the subsequent Zymo-Spin™ IICR Column efficiently binds total RNA. RNA is eluted in 25 l and is suitable for subsequent procedures including RT-qPCR
Catalog No. 50-444-617 Supplier Zymo Research Corporation Supplier No. R2014
Ultra-high density ZR BashingBeads™ are included for sample homogenization coupled with a robust buffer system, delivering total RNA (including small/microRNAs).The Zymo-Spin™ IIICG Column allows for high-capacity DNA elimination and the subsequent Zymo-Spin™ IICR Column efficiently binds total RNA. RNA is eluted in 25 l and is suitable for subsequent procedures including RT-qPCR