Learn More
Zymo Research Corporation Quick-RNA Plant MiniPrep™ Kit (50 Preps)
SDP
Catalog No. 50444618
Encompass_Preferred
The kit includes unique technology such as the ZR BashingBead™ Lysis Tubes and features a specially formulated RNA Lysis Buffer. The Zymo-Spin™ IIICG Column allows for high-capacity DNA elimination and the subsequent Zymo-Spin™ IICR Column efficiently adsorbs total RNA.The RNA is washed and then eluted with DNase/RNase-Free Water. For inhibitor removal, the eluted RNA can be treated by running the sample through the Zymo-Spin™ III-HRC Filters. RNA is eluted in as little as 6 l and is suitable for subsequent procedures including RT-qPCR.
Catalog No. 50-444-618 Supplier Zymo Research Corporation Supplier No. R2024
The kit includes unique technology such as the ZR BashingBead™ Lysis Tubes and features a specially formulated RNA Lysis Buffer. The Zymo-Spin™ IIICG Column allows for high-capacity DNA elimination and the subsequent Zymo-Spin™ IICR Column efficiently adsorbs total RNA.The RNA is washed and then eluted with DNase/RNase-Free Water. For inhibitor removal, the eluted RNA can be treated by running the sample through the Zymo-Spin™ III-HRC Filters. RNA is eluted in as little as 6 l and is suitable for subsequent procedures including RT-qPCR.