Learn More
Zymo Research Corporation ZR RNA Buffer, 50 ml
SDP
Catalog No. 502037232
Encompass_Preferred
ZR RNA Buffer is a one-step lysis and binding buffer for the purification of RNA in the Quick-RNA Midiprep Kit.
Catalog No. 50-203-7232 Supplier Zymo Research Corporation Supplier No. R1020150
ZR RNA Buffer is a one-step lysis and binding buffer for the purification of RNA in the Quick-RNA Midiprep Kit.