Learn More
Zymo Research Corporation ZR small-RNA™ Ladder (10 μg)
SDP
Catalog No. 50444611
Encompass_Preferred
The ladder consists of four single-stranded RNA oligonucleotides 17, 21, 25, and 29 bases in length. The marker is supplied in water and can be stained with dyes specific for single-stranded nucleic acid species e.g, GelStar.
Catalog No. 50-444-611 Supplier Zymo Research Corporation Supplier No. R1090
The ladder consists of four single-stranded RNA oligonucleotides 17, 21, 25, and 29 bases in length. The marker is supplied in water and can be stained with dyes specific for single-stranded nucleic acid species e.g, GelStar.