Learn More
Zymo Research Corporation ZR small-RNA™ PAGE Recovery Kit (20 Preps)
SDP
Catalog No. 50444607
Encompass_Preferred
The ZR small-RNA™ PAGE Recovery Kit is a refinement of the crush & soak method that incorporates a unique buffer system together with Zymo-Spin™ Column technology for improved recovery and added convenience. The recovered RNA can be concentrated at elution step in volumes as small as 6 l and is ideal for any downstream enzymatic reaction or manipulation.
Catalog No. 50-444-607 Supplier Zymo Research Corporation Supplier No. R1070
The ZR small-RNA™ PAGE Recovery Kit is a refinement of the crush & soak method that incorporates a unique buffer system together with Zymo-Spin™ Column technology for improved recovery and added convenience. The recovered RNA can be concentrated at elution step in volumes as small as 6 l and is ideal for any downstream enzymatic reaction or manipulation.