Learn More
Zymo Research Corporation ZR Urine RNA Isolation™ Kit (20 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. 50444584
Encompass_Preferred
Cells from urine are separated using a syringe (not provided) and a uniquely-designed syringe filter. Cells are then lysed and total RNA is stabilized using a specially formulated Urine RNA Buffer. The collected lysate can then be used immediately or at a later time following transportation and/or storage. Also, this kit is ideal for direct isolation of RNA from microvesicles that may be recovered from urine filtrates. One-step RNA isolation occurs via matrix adsorption using Zymo-Spin™ IC Columns, then washed and eluted. High-quality, total RNA from urine samples is suitable for subsequent analyses of gene expression that include NGS, RT/qPCR, etc.
Catalog No. 50-444-584 Supplier Zymo Research Corporation Supplier No. R1038
Add to Cart
Add to Cart
Cells from urine are separated using a syringe (not provided) and a uniquely-designed syringe filter. Cells are then lysed and total RNA is stabilized using a specially formulated Urine RNA Buffer. The collected lysate can then be used immediately or at a later time following transportation and/or storage. Also, this kit is ideal for direct isolation of RNA from microvesicles that may be recovered from urine filtrates. One-step RNA isolation occurs via matrix adsorption using Zymo-Spin™ IC Columns, then washed and eluted. High-quality, total RNA from urine samples is suitable for subsequent analyses of gene expression that include NGS, RT/qPCR, etc.