Learn More
Zymo Research Corporation Quick-DNA™ Viral Kit (50 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Supplier:  Zymo Research Corporation D3015
Encompass_Preferred
Quick recovery of viral DNA from a wide range of sources using Zymo-Spin column technology. Column design allows DNA to be eluted at high concentrations into minimal volumes. Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion.
Catalog No. 50-444-141