Learn More
Research Products International Corp ZR Viral RNA Kit w/Zymo-Spin IC Columns, Capped, 50 Preps
SDP
Supplier:  Research Products International Corp ZR1034
Encompass
The Quick-RNA Viral Kit is a quick viral RNA purification kit for viral RNA from plasma, serum, cell culture media, cellular suspensions, urine, blood, saliva and any other biological samples stored in DNA/RNA Shield. DNA/RNA Shield ensures nucleic acid stability during sample storage/transport at ambient temperatures (4-25°C). The reagent effectively lyses cells and inactivates nucleases and infectious agents (virus). The kit also features a specialized buffer system that facilitates complete viral particle lysis for efficient RNA isolation from samples containing enteroviruses, rhinoviruses, coronaviruses, HIV, HCV, influenza A virus, flaviviruses, measles virus, parainfluenza virus, parvovirus (a ssDNA virus), etc. Viral RNA is bound to the column, washed and eluted.
Catalog No. 50-197-7625